The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006140.2 | NP_001006140.2 | 3 - 4 | 289 | 93 |
GenBank mRNA | AJ720805.1 | - | 3 - 4 | 290 | 93 |