Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC174
Entrez Gene ID
416044
Gene Name
coiled-coil domain containing 174
Gene Aliases
C12H3ORF19, C3orf19
Chromosome Location
Chr.12: 11645040 - 11656384 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006140.2 NP_001006140.2 6 - 7 627 127
GenBank mRNA AJ720805.1 - 6 - 7 628 127

View Full Product Details