Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP4R2
Entrez Gene ID
416098
Gene Name
protein phosphatase 4 regulatory subunit 2
Gene Aliases
-
Chromosome Location
Chr.12: 16915947 - 16950996 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006142.1 NP_001006142.1 7 - 8 788 93
XM_025154495.1 XP_025010263.1 10 - 11 6541 93
GenBank mRNA AJ719288.1 - 7 - 8 788 93

View Full Product Details