Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APMAP
Entrez Gene ID
416715
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
BSCv, C20orf3, C3H20ORF3
Chromosome Location
Chr.3: 15927402 - 15940460 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006177.1 NP_001006177.1 8 - 9 1050 113
GenBank mRNA AJ720833.1 - 8 - 9 1050 113
CR389678.1 - 8 - 9 1049 113

View Full Product Details