Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF34
Entrez Gene ID
416843
Gene Name
ring finger protein 34
Gene Aliases
-
Chromosome Location
Chr.15: 5762349 - 5771343 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006188.1 NP_001006188.1 5 - 6 930 65
XM_015275281.2 XP_015130767.1 5 - 6 1004 65
XM_025155454.1 XP_025011222.1 5 - 6 909 65
GenBank mRNA AJ719798.1 - 5 - 6 930 65
BX933911.2 - 5 - 6 912 65

View Full Product Details