Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POC1B
Entrez Gene ID
417889
Gene Name
POC1 centriolar protein B
Gene Aliases
WDR51B
Chromosome Location
Chr.1: 43360801 - 43404429 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006232.1 NP_001006232.1 6 - 7 783 74
GenBank mRNA AJ719544.1 - 6 - 7 783 74

View Full Product Details