Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H21ORF59
Entrez Gene ID
418497
Gene Name
chromosome 1 open reading frame, human C21orf59
Gene Aliases
C21orf59
Chromosome Location
Chr.1: 106340922 - 106348250 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006258.1 NP_001006258.1 4 - 5 605 142
GenBank mRNA AJ720779.1 - 4 - 5 605 142

View Full Product Details