Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB9
Entrez Gene ID
418625
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
ASB-9
Chromosome Location
Chr.1: 123496141 - 123510438 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006262.1 NP_001006262.1 6 - 7 686 55
XM_015273215.2 XP_015128701.1 5 - 6 746 55
XM_015273223.2 XP_015128709.1 6 - 7 919 55
GenBank mRNA AJ720472.1 - 6 - 7 686 55

View Full Product Details