Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UQCC1
Entrez Gene ID
419139
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 1
Gene Aliases
UQCC
Chromosome Location
Chr.20: 1275269 - 1328733 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006285.1 NP_001006285.1 3 - 4 246 81
XM_015296246.2 XP_015151732.1 3 - 4 348 81
XM_025142189.1 XP_024997957.1 3 - 4 534 81
GenBank mRNA AJ720641.1 - 3 - 4 246 81

View Full Product Details