Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP4B
Entrez Gene ID
419150
Gene Name
charged multivesicular body protein 4B
Gene Aliases
-
Chromosome Location
Chr.20: 2120742 - 2142286 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006286.1 NP_001006286.1 2 - 3 564 106
GenBank mRNA AJ721089.1 - 2 - 3 564 106

View Full Product Details