Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELFCD
Entrez Gene ID
419312
Gene Name
negative elongation factor complex member C/D
Gene Aliases
TH1-like, TH1L
Chromosome Location
Chr.20: 10901830 - 10910619 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006294.1 NP_001006294.1 13 - 14 1626 92
GenBank mRNA AJ720726.1 - 13 - 14 1626 92

View Full Product Details