Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAPB
Entrez Gene ID
419317
Gene Name
VAMP associated protein B and C
Gene Aliases
-
Chromosome Location
Chr.20: 11174597 - 11208289 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006296.1 NP_001006296.1 4 - 5 430 110
GenBank mRNA AJ720441.1 - 4 - 5 430 110

View Full Product Details