Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTFDC1
Entrez Gene ID
419329
Gene Name
replication termination factor 2 domain containing 1
Gene Aliases
C20H20ORF43, C20orf43, CDAO5, HSPC164, RTF2, SHUJUN-3
Chromosome Location
Chr.20: 11969388 - 11990677 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006297.2 NP_001006297.2 5 - 6 534 146
XM_015296411.2 XP_015151897.1 5 - 6 526 146
GenBank mRNA AJ720403.1 - 5 - 6 537 146

View Full Product Details