Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCK
Entrez Gene ID
422648
Gene Name
deoxycytidine kinase
Gene Aliases
-
Chromosome Location
Chr.4: 50129676 - 50140113 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006451.1 NP_001006451.1 6 - 7 817 96
GenBank mRNA AJ719431.1 - 6 - 7 817 96

View Full Product Details