Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H15
Entrez Gene ID
423992
Gene Name
zinc finger CCCH-type containing 15
Gene Aliases
DFRP1, GDFRP1
Chromosome Location
Chr.7: 1406698 - 1418831 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006510.1 NP_001006510.1 6 - 7 853 156
GenBank mRNA AB185935.1 - 6 - 7 837 156
AJ720056.1 - 6 - 7 853 156

View Full Product Details