Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNOT9
Entrez Gene ID
424215
Gene Name
CCR4-NOT transcription complex subunit 9
Gene Aliases
RQCD1
Chromosome Location
Chr.7: 22491554 - 22503173 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006521.2 NP_001006521.2 5 - 6 610 82
XM_015289800.2 XP_015145286.1 5 - 6 624 82
GenBank mRNA AJ720224.1 - 5 - 6 610 82

View Full Product Details