Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPH2
Entrez Gene ID
424574
Gene Name
DPH2 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 20715238 - 20717610 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006538.1 NP_001006538.1 1 - 2 179 111
GenBank mRNA AJ720102.1 - 1 - 2 179 111

View Full Product Details