Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYG1
Entrez Gene ID
425050
Gene Name
glycogenin 1
Gene Aliases
GYG, glycogenin-1
Chromosome Location
Chr.9: 24096063 - 24111184 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006558.1 NP_001006558.1 2 - 3 209 94
XM_015291687.2 XP_015147173.1 2 - 3 215 94
XM_015291688.2 XP_015147174.1 3 - 4 301 94
XM_015291689.2 XP_015147175.1 3 - 4 274 94
XM_015291691.2 XP_015147177.1 2 - 3 238 94
XM_015291692.2 XP_015147178.1 2 - 3 215 94
XM_025153395.1 XP_025009163.1 2 - 3 215 94
GenBank mRNA AJ720384.1 - 2 - 3 209 94

View Full Product Details