Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYG1
Entrez Gene ID
425050
Gene Name
glycogenin 1
Gene Aliases
GYG, glycogenin-1
Chromosome Location
Chr.9: 24096063 - 24111184 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006558.1 NP_001006558.1 6 - 7 898 120
XM_015291688.2 XP_015147174.1 7 - 8 990 120
XM_015291689.2 XP_015147175.1 7 - 8 963 120
XM_015291691.2 XP_015147177.1 6 - 7 927 120
GenBank mRNA AJ720384.1 - 6 - 7 898 120

View Full Product Details