Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA7
Entrez Gene ID
426787
Gene Name
golgin A7
Gene Aliases
-
Chromosome Location
Chr.22: 2899324 - 2903252 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006570.1 NP_001006570.1 4 - 5 624 112
GenBank mRNA AJ719805.1 - 4 - 5 624 112
CR387916.1 - 4 - 5 510 112

View Full Product Details