Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRX1
Entrez Gene ID
373941
Gene Name
paired related homeobox 1
Gene Aliases
PMX1, Prx-1, Prx1, gMHox
Chromosome Location
Chr.8: 5216524 - 5248945 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277724.2 NP_001264653.1 1 - 2 478 112
XM_015290230.2 XP_015145716.1 1 - 2 565 112
XM_015290231.2 XP_015145717.1 1 - 2 564 112
GenBank mRNA AF461698.1 - 1 - 2 294 112
AL585199.1 - 1 - 2 481 112
BU311875.1 - 1 - 2 65 112
BX258858.4 - 1 - 2 400 112
CD214940.1 - 1 - 2 478 112
D13433.1 - 1 - 2 475 112
S69088.1 - 1 - 2 273 112

View Full Product Details