Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLOC1S6
Entrez Gene ID
415447
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 6
Gene Aliases
PLDN, pallidin
Chromosome Location
Chr.10: 11145530 - 11151303 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007825.1 NP_001007826.1 1 - 2 156 88
GenBank mRNA AJ720762.1 - 1 - 2 156 88

View Full Product Details