Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGFR1OP2
Entrez Gene ID
418214
Gene Name
FGFR1 oncogene partner 2
Gene Aliases
-
Chromosome Location
Chr.1: 67983452 - 67994763 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007855.1 NP_001007856.1 5 - 6 575 93
XM_015290277.2 XP_015145763.1 5 - 6 622 93
GenBank mRNA AJ720090.1 - 5 - 6 575 93

View Full Product Details