Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP2
Entrez Gene ID
422678
Gene Name
fatty acid binding protein 2
Gene Aliases
FABP
Chromosome Location
Chr.4: 54716756 - 54719201 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007923.1 NP_001007924.1 1 - 2 65 109
GenBank mRNA BX930518.1 - 1 - 2 88 109
BX932340.2 - 1 - 2 82 109
BX933597.1 - 1 - 2 85 109

View Full Product Details