Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCM2
Entrez Gene ID
428478
Gene Name
glial cells missing homolog 2
Gene Aliases
-
Chromosome Location
Chr.2: 63211417 - 63216473 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008480.2 NP_001008480.1 3 - 4 424 76
GenBank mRNA AB175672.1 - 3 - 4 425 76

View Full Product Details