Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPARD
Entrez Gene ID
395479
Gene Name
peroxisome proliferator activated receptor delta
Gene Aliases
PPARBETA
Chromosome Location
Chr.26: 4087839 - 4106014 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204728.1 NP_990059.1 - 630 50
XM_015298888.2 XP_015154374.1 - 829 50
XM_015298889.2 XP_015154375.1 - 1077 50
XM_015298890.2 XP_015154376.1 - 554 50
XM_015298891.2 XP_015154377.1 - 816 50
XM_015298892.2 XP_015154378.1 - 2477 50
XM_015298893.2 XP_015154379.1 - 677 50
XM_025143663.1 XP_024999431.1 - 619 50
XM_025143664.1 XP_024999432.1 - 890 50
XM_025143665.1 XP_024999433.1 - 919 50
XM_025143666.1 XP_024999434.1 - 767 50
XM_025143667.1 XP_024999435.1 - 1199 50
XM_025143668.1 XP_024999436.1 - 623 50
GenBank mRNA AF163810.1 - - 630 50

View Full Product Details