Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNB
Entrez Gene ID
395515
Gene Name
taxilin beta
Gene Aliases
MDP77
Chromosome Location
Chr.3: 54077243 - 54115446 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204754.1 NP_990085.1 9 - 10 1457 93
XM_015284009.2 XP_015139495.1 9 - 10 1487 93
XM_025148524.1 XP_025004292.1 9 - 10 1431 93
GenBank mRNA D89999.1 - 9 - 10 1457 93

View Full Product Details