Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT4
Entrez Gene ID
395561
Gene Name
Wnt family member 4
Gene Aliases
-
Chromosome Location
Chr.21: 6313309 - 6325046 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204783.1 NP_990114.1 1 - 2 264 73
GenBank mRNA D31900.1 - 1 - 2 264 73

View Full Product Details