Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT11
Entrez Gene ID
395562
Gene Name
Wnt family member 11
Gene Aliases
-
Chromosome Location
Chr.1: 195311154 - 195346046 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204784.1 NP_990115.1 2 - 3 433 70
XM_015280851.2 XP_015136337.1 3 - 4 1322 70
XM_015280852.2 XP_015136338.1 3 - 4 1322 70
GenBank mRNA D31901.1 - 2 - 3 433 70

View Full Product Details