Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCXD1
Entrez Gene ID
418673
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 131370555 - 131388802 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128637.1 NP_001122109.1 6 - 7 773 82
XM_015277705.2 XP_015133191.1 6 - 7 817 82
XM_015277706.2 XP_015133192.1 6 - 7 843 82

View Full Product Details