Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPCN3
Entrez Gene ID
421224
Gene Name
two-pore calcium channel 3
Gene Aliases
-
Chromosome Location
Chr.3: 3092562 - 3106764 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143931.1 NP_001137403.1 18 - 19 2111 93
XM_015283362.2 XP_015138848.1 17 - 18 1942 93
GenBank mRNA EU344154.1 - 18 - 19 2111 93

View Full Product Details