Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMP3
Entrez Gene ID
424961
Gene Name
lysosomal associated membrane protein 3
Gene Aliases
DC-LAMP
Chromosome Location
Chr.9: 16528555 - 16540200 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146132.1 NP_001139604.1 1 - 2 49 70
GenBank mRNA AM933592.1 - 1 - 2 49 70

View Full Product Details