The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146133.1 | NP_001139605.1 | 4 - 4 | 752 | 80 |
GenBank mRNA | EU282004.1 | - | 4 - 4 | 752 | 80 |