Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MARCKS
Entrez Gene ID
396473
Gene Name
myristoylated alanine rich protein kinase C substrate
Gene Aliases
-
Chromosome Location
Chr.3: 64897927 - 64902679 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205480.1 NP_990811.1 2 - 2 1349 95
XM_015284506.2 XP_015139992.1 3 - 3 2140 95
GenBank mRNA CR354248.1 - 1 - 1 746 95
M31650.1 - 2 - 2 1349 95

View Full Product Details