Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPZ1
Entrez Gene ID
547466
Gene Name
nonclathrin coat protein zeta1-COP
Gene Aliases
-
Chromosome Location
Chr.7: 580451 - 583800 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249958.1 NP_001236887.1 2 - 3 231 118
GenBank mRNA AB040040.1 - 2 - 3 216 118
BT089454.1 - 2 - 3 231 118

View Full Product Details