Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GH1
- Entrez Gene ID
-
547502
- Gene Name
- GH1 protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 1886333 - 1890391 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF016633.1 |
- |
3 - 4 |
772 |
64 |
- Gene Symbol
- LOC100785203
- Entrez Gene ID
-
100785203
- Gene Name
- auxin-responsive protein IAA8-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 40063560 - 40067914 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BT095721.1 |
- |
4 - 5 |
868 |
64 |
View Full Product Details