Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN1
Entrez Gene ID
547540
Gene Name
mannosyl-oligosaccharide 1,2-alpha-mannosidase
Gene Aliases
-
Chromosome Location
Chr.8: 18078729 - 18085367 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251128.1 NP_001238057.1 4 - 5 858 92
GenBank mRNA AF126550.1 - 4 - 5 858 92

View Full Product Details