Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAE1
Entrez Gene ID
547598
Gene Name
20S proteasome subunit
Gene Aliases
-
Chromosome Location
Chr.10: 1283088 - 1287371 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001248230.1 NP_001235159.1 4 - 5 305 69
GenBank mRNA AF255338.1 - 4 - 5 305 69

View Full Product Details