Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBP2
Entrez Gene ID
548002
Gene Name
sucrose-binding protein 2
Gene Aliases
SBP
Chromosome Location
Chr.2: 14850163 - 14853756 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250315.1 NP_001237244.1 3 - 4 752 96
GenBank mRNA AF191299.1 - 3 - 4 769 96
AK244143.1 - 3 - 4 787 96
AY234869.1 - 3 - 4 752 96
BT093571.1 - 3 - 4 788 96

View Full Product Details