The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251224.1 | NP_001238153.1 | 11 - 12 | 1344 | 69 |
GenBank mRNA | AY492005.1 | - | 11 - 12 | 1344 | 69 |
FN564570.1 | - | 11 - 12 | 1344 | 69 | |
FN564571.1 | - | 11 - 12 | 1344 | 69 |