Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DGD1
- Entrez Gene ID
-
548059
- Gene Name
- digalactosyldiacylglycerol synthase 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 45583946 - 45589596 on Build Glycine_max_v2.1
- Gene Symbol
- LOC100796354
- Entrez Gene ID
-
100796354
- Gene Name
- digalactosyldiacylglycerol synthase 1, chloroplastic-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 43046534 - 43052196 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK287394.1 |
- |
5 - 6 |
1487 |
85 |
View Full Product Details