Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDS1
- Entrez Gene ID
-
547970
- Gene Name
- phytoene desaturase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 317379 - 325683 on Build Glycine_max_v2.1
- Gene Symbol
- LOC100127420
- Entrez Gene ID
-
100127420
- Gene Name
- choloroplast phytoene desaturase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 38140862 - 38147805 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK244738.1 |
- |
3 - 4 |
526 |
61 |
View Full Product Details