Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAS3
Entrez Gene ID
732660
Gene Name
asparagine synthetase, type III
Gene Aliases
-
Chromosome Location
Chr.18: 1223195 - 1229489 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AM158274.1 - 10 - 11 1391 76

View Full Product Details