Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSH1
Entrez Gene ID
732638
Gene Name
DNA mismatch repair protein
Gene Aliases
-
Chromosome Location
Chr.10: 34811698 - 34849624 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251288.1 NP_001238217.1 20 - 21 2568 104
GenBank mRNA AY856369.1 - 20 - 21 2568 104

View Full Product Details