Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLS1
Entrez Gene ID
778157
Gene Name
flavonol synthase
Gene Aliases
gmfls1
Chromosome Location
Chr.13: 2707847 - 2712767 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250490.1 NP_001237419.1 1 - 2 493 74
GenBank mRNA AB246668.1 - 1 - 2 493 74
AK286832.1 - 1 - 2 508 74

View Full Product Details