Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY17
Entrez Gene ID
100127373
Gene Name
WRKY17 protein
Gene Aliases
-
Chromosome Location
Chr.6: 4658795 - 4660526 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254057.2 NP_001240986.1 2 - 3 511 82
GenBank mRNA BT095976.1 - 2 - 3 513 82
EU019561.1 - 1 - 2 124 82

View Full Product Details