Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPK1
Entrez Gene ID
100127406
Gene Name
inositol pentakisphosphate 2-kinase
Gene Aliases
-
Chromosome Location
Chr.14: 5949463 - 5952353 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250504.1 NP_001237433.1 2 - 3 316 99
GenBank mRNA EU033956.1 - 2 - 3 316 99
KF359958.1 - 2 - 3 316 99

View Full Product Details