Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY4
Entrez Gene ID
100170684
Gene Name
transcription factor
Gene Aliases
-
Chromosome Location
Chr.8: 20589387 - 20593841 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250726.1 NP_001237655.1 1 - 2 190 63
GenBank mRNA EU375355.1 - 1 - 2 190 63

View Full Product Details