Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNK10
Entrez Gene ID
100272201
Gene Name
with no lysine kinase 10
Gene Aliases
-
Chromosome Location
Chr.9: 45963410 - 45967330 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249075.1 NP_001236004.1 6 - 7 1442 71
GenBank mRNA FJ795040.1 - 6 - 7 1442 71

View Full Product Details