Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY55
Entrez Gene ID
100127428
Gene Name
WRKY55 protein
Gene Aliases
-
Chromosome Location
Chr.7: 5052692 - 5055577 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001255746.1 NP_001242675.1 3 - 3 1595 102
GenBank mRNA BT093956.1 - 3 - 3 1595 102
EU019588.1 - 3 - 3 950 102

View Full Product Details